Learn More
Brady™ BMP53 Label Printer Kit
Product Code. 11932732
Change view
Click to view available options
Software Type:
LabelMark
LabelMark, CodeSoft
None
Unit Size:
Each
Product Code. Software Type unitSize
11932732 None Each
11922732 LabelMark Each
12673910 LabelMark, CodeSoft Each
3 options
Product Code. 11932732
Brand: Brady™ 710895
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Label Printer EU
Print Technology Thermal Transfer
Width (Metric) 165 mm
Height (Metric) 90 mm
Depth (Metric) 215 mm
Dimensions (L x W) 38.1 mm W
Color Capability Monochrome
Includes Li-ion battery, USB cable, AC adapter with power cord, Product CD, CD with Brady printer drivers, Quick Start Guide, Bluetooth network card and MC-1500-595-WT-BK
Model BMP53
Print Speed 25.4 mm/sec.
Resolution 300 dpi
Software Type None
Connector Types USB, Bluetooth. Optional: Ethernet, WiFi
Weight (Metric) 1.28 kg
For Use With (Application) For labeling various purposes, such as electric field, data communication, safety and building, product and panel manufacturing, and laboratories
Quantity 1 Ea.
Warranty 2 Years
Power Supply Details Battery Pack, AC Adaptor/Battery Charger