Learn More
Brady™ BMP51 Laboratory Kit for EU
The BMP51 Label Maker is a compact, portable printer with a built-in cutter and easy top-loading cartridges. Ideal for producing professional laboratory labels.
Brand: Brady™ 198655
9511.88 NOK valid until 2024-06-30
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 5.76100kg Transport : UN number : 3481 Chem class : 9
Description
When you need clear, durable labels on the spot, you need the BMP51 Label Printer from Brady™. It offers crisp printing, easy formatting, convenient label sizes, and portability required for electrical and datacom installations, laboratory identification, facility and safety labeling. This kit comes with the Brady Workstation LAB suite, enabling you to immediately start designing professional labels.
- Prints durable labels that perform throughout sample storage and processing
- Change supplies in seconds with drop-in cartridges that have label material and ribbon inside
- Cartridges feature smart cell technology that automatically formats your label correctly
- Prints a speedy 25.4 mm/s, meaning no more waiting around for labels to print
- Use with AC power supply, AA batteries, or the rechargeable Lithium-Ion battery which can print over 3000 labels on a single charge
- Compatible with mobile apps
- Full QWERTY keyboard
- Built-in Bluetooth™ technology and optional Wi-Fi card provide wireless printing options
Specifications
Blue | |
Thermal Transfer | |
155 mm | |
262 mm | |
91 mm | |
Non-sterile | |
Printer, Li-ion Battery, USB, AC Adaptor with EU Power Cord, Product CD, Brady™ Printer Drivers CD, Quick Start Guide, Bluetooth™ Network Card, Brady™ Workstation LAB Suite, Cartridge | |
48 MB | |
25.4 mm/s | |
Bluetooth™, Ethernet, USB | |
2.6 lb. | |
2011/65/EU and 2015/863 | |
2 Year |
Monocolor LCD | |
6 in. | |
10.3 in. | |
3.6 in. | |
Not Autoclavable | |
Monochrome | |
Danish, Swedish, English, English, French, French, Italian, Italian, Portuguese, Spanish, Spanish | |
BMP51 | |
300 dpi | |
4°C to 43°C | |
1.2 kg | |
Data Labeling, Electrical and Laboratory Labeling, General Identification | |
110/240 V |