Learn More
Brady™ BMP™71 Series WorkHorse™ Permanent Polyester Labels - White
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand: Brady™ 114775
Additional Details : Weight : 0.08100kg
Description
- Glossy white finish with permanent acrylic is thermal transfer printable
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
- UL approved
- CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Not Autoclavable | |
Label | |
White | |
Polyester | |
Label | |
1 | |
100 Labels/Roll | |
2 x 1 in. (W x H) | |
2 in. | |
130.81 mm | |
1 in. |
ASTM B 117; ASTM D 1000; ASTM D 2979; ASTM G 155; CSA Approved; Halogen Free (DIN VDE 0472 part 815); UL Recognized | |
Permanent Acrylic | |
Roll | |
Thermal Transfer | |
BMP™71 Label Printer | |
No Text | |
100/Roll | |
1 x 2 in. | |
2 in. | |
2 in. | |
1 in. |