Learn More
Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels
Click to view available options
For Use With (Equipment):
BMP41, BMP51, BMP53 printers
BMP51 and BMP53 Printer
Size (English):
0.375 in. x 20 ft.
0.5 x 1 in.
0.75 x 0.5 in.
1 x 1.25 in.
1 x 1.9 in.
Includes:
1 (cont.)
100 labels
160 labels
180 labels
220 labels
Unit Size:
Each
Description
Designed for use with various BMP™50 series label printers. Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels offer a repositionable adhesive and are intended for use in wire and cable marking, component or equipment identification, barcoding, and other general identification.
- Adhesive and cloth backing offers excellent holding power, yet allow for clean removal and repositioning
- Stick to any clean, dry surface
- Specially formulated topcoat for excellent print quality
- All-in-one cartridge includes label supply and R6200 Series ribbon
Specifications
Specifications
| Certifications/Compliance | RoHS, UL |
| Description | BMP41/BMP51/BMP53 Labelmaker Labels |
| Adhesive Type | Rubber, Repositionable |
| Color | Black, White |
| Finish | Semi-gloss |
| Format | Cartridge |
| Material | Vinyl Cloth (B-498) |
| Print Technology | Thermal Transfer |
| Display Text | None |
| For Use With (Application) | Barcoding, Component ID, Equipment ID, General purpose ID, Wire and cable marking |
| Show More |