missing translation for 'onlineSavingsMsg'
Learn More

Bmf Antibody, Novus Biologicals™

Product Code. 18645022 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
Product Code. Quantity unitSize
18645022 100 μg 100µL
18669524 50 μg 50µL
2 options
This item is not returnable. View return policy

Product Code. 18645022

Brand: Novus Biologicals NBP297262100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Bmf Polyclonal antibody specifically detects Bmf in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bmf
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.0)
Gene Alias Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Bmf fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 90427
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.