Learn More
Melford Bluo-Gal
Product Code. 30018107
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
Product Code. 30018107
Brand: Melford B725000.1
Chromogenic substrate for B-D-Galactosidase.
Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.