Learn More
Sampling Systems™ Blue SteriWare™ Liquid Funnels
Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Description
- Our range of single use, disposable liquid funnels are ideal for use in laboratories and manufacturing areas.
- These funnels are made from food grade polymers and are designed to be used straight from the bag.
- The bright blue color means they can be easily seen if they drop in or left in the material by mistake.
- SteriWare™ Funnels are available sterile and non-sterile.
- SteriWare™ is the largest range of single-use equipment for sampling, laboratory and production work on the market.
- The whole range is made in a cleanroom and comes with class-leading pharmaceutical documentation.
- Simply unwrap the funnel and use.
- The funnels come with a side tab so they are easy to hold.
- The individually bagged funnels are made from PP that is FDA/EC compliant.
- The material used is also certified BSE/TSE free. The sterile funnels conform to SAL 10-6
- The funnels come with a batch-specific certificate that includes the date of manufacture, expiry date and irradiation certificate (for sterile funnels only).
Specifications
Specifications
| Diameter (Metric)Top | 100 mm |
| Material | Polypropylene |
| Color | Blue |
| Sterility | Sterile |