Learn More
Keystone Cleanroom Products Blue SMS Sterilization Wrap, 50 gsm
BIOPHARMAGREENER_CHOICE
Product Code. 17861481 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
10.2 x 10.2 cm (4 x 4 in.)
101.6 x 101.6 cm (40 x 40 in.)
114.3 x 114.3 cm (45 x 45 in.)
114.3 x 121.9 cm (45 x 48 in.)
121.9 x 121.9 cm (48 x 48 in.)
137.2 x 137.2 cm (54 x 54 in.)
137.2 x 182.9 cm (54 x 72 in.)
15.2 x 15.2 cm (6 x 6 in.)
152.4 x 152.4 cm (60 x 60 in.)
152.4 x 203.2 cm (60 x 80 in.)
20.3 x 20.3 cm (8 x 8 in.)
22.9 x 22.9 cm (9 x 9 in.)
22.9 x 30.5 cm (9 x 12 in.)
3.8 x 3.8 cm (1.5 x 1.5 in.)
30.5 x 30.5 cm (12 x 12 in.)
38.1 x 38.1 cm (15 x 15 in.)
45.7 x 45.7 cm (18 x 18 in.)
45.7 x 55.9 cm (18 x 22 in.)
5.1 x 5.1 cm (2 x 2 in.)
50.8 x 50.8 cm (20 x 20 in.)
61 x 61 cm (24 x 24 in.)
7.6 x 7.6 cm (3 x 3 in.)
76.2 x 76.2 cm (30 x 30 in.)
91.4 x 91.4 cm (36 x 36 in.)
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Pack of 3000
Pack of 50
Product Code. Dimensions (L x W) unitSize
17861481 45.7 x 55.9 cm (18 x 22 in.) Pack of 1000
17851471 3.8 x 3.8 cm (1.5 x 1.5 in.) Pack of 3000
17861471 5.1 x 5.1 cm (2 x 2 in.) Pack of 3000
17871471 7.6 x 7.6 cm (3 x 3 in.) Pack of 3000
17881471 10.2 x 10.2 cm (4 x 4 in.) Pack of 3000
17891471 15.2 x 15.2 cm (6 x 6 in.) Pack of 3000
17801481 20.3 x 20.3 cm (8 x 8 in.) Pack of 1000
17811481 22.9 x 22.9 cm (9 x 9 in.) Pack of 1000
17821481 22.9 x 30.5 cm (9 x 12 in.) Pack of 1000
17831481 30.5 x 30.5 cm (12 x 12 in.) Pack of 1000
17841481 38.1 x 38.1 cm (15 x 15 in.) Pack of 1000
17851481 45.7 x 45.7 cm (18 x 18 in.) Pack of 1000
17871481 50.8 x 50.8 cm (20 x 20 in.) Pack of 1000
17881481 61 x 61 cm (24 x 24 in.) Pack of 250
17891481 76.2 x 76.2 cm (30 x 30 in.) Pack of 250
17801491 91.4 x 91.4 cm (36 x 36 in.) Pack of 250
17811491 101.6 x 101.6 cm (40 x 40 in.) Pack of 250
17821491 114.3 x 114.3 cm (45 x 45 in.) Pack of 250
17831491 114.3 x 121.9 cm (45 x 48 in.) Pack of 250
17841491 121.9 x 121.9 cm (48 x 48 in.) Pack of 250
17851491 137.2 x 137.2 cm (54 x 54 in.) Pack of 100
17861491 137.2 x 182.9 cm (54 x 72 in.) Pack of 100
17871491 152.4 x 152.4 cm (60 x 60 in.) Pack of 100
17881491 152.4 x 203.2 cm (60 x 80 in.) Pack of 50
24 options
Product Code. 17861481
Brand: Keystone Cleanroom Products KCRP3001822
Only null left
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
SMS sterilization wraps provide a microbial barrier and are ideal for pharmaceutical production applications. They are designed for use in steam, ethylene oxide, and hydrogen peroxide sterilization processes.
  • Manufactured and packaged in a Certified ISO 8 cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm material
  • Soft and drape-able fabric
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gasses but prevents liquid and microbial penetration
  • Sub packed into sealed labeled poly bags with Lot Number and Date of Manufacture for complete traceability
  • Certificate of Conformance sent with each shipment
  • Made in the USA
Spezifikation
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type None
Color Blue
Disposable Single-use
Material SMS Fabric
Sterility Non-sterile
Latex Free Yes
Temperature Range (Metric) 121°C to 125°C
Dimensions (L x W) 45.7 x 55.9 cm (18 x 22 in.)
Length (English) 22 in.
Length (Metric) 55.9 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Product Type Sterilization Wrap (Heavy-duty)
Mehr anzeigen Weniger anzeigen