Learn More
Syngene™ Blue Slimline LED Transilluminator with Sliding Tray
Product Code. 16574331 Shop All Syngene Products
Change view
Click to view available options
Wavelength:
470 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
16574331 470 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16574331 Supplier Syngene™ Supplier No. GXULTRASLIMLED
  • ONLINE EXCLUSIVE PRICE
    10490.00 NOK
    9846.51 NOK / Pack of 1
    Save 643.49 NOK
    6% Off
 Restricted Item
Estimated Shipment: 27-01-2027
LED Transilluminator suitable for a variety of imaging needs.
  • Even blue epi LED illumination
  • High sensitivity with improved S/N ratio
  • Image a range of fluorophores- EtBr, SYBRSafe, GelGreen, SYBR Gold, SYBRGreen, SYPRORuby, SYPROOrange, GelRed
  • LED is safe to skin and eyes with no damage to DNA
Specifications
Product Type Transilluminator
Includes Sliding tray
Size (Metric) Filter 7 x 12 cm
Wavelength 470 nm