Learn More
Scotch-Brite™ Blue Floor Pads
Description
- Heavy-duty scrub pad removes dirt, spills, scuffs, and 1-2 coats of finish
- Leaves surfaces clean for recoating
- Designed for frequent burnishing with less dust and burning
- Washable and reusable pad is economical and helps postpone end-of-life disposal to landfills
- Versatile pads can be used for heavy-duty spray cleaning and foam scrubbing
- Available in sizes for rotary and automatic floor machines running at low speed (175-600 rpm)
Specifications
Specifications
| Product Type | Stripping Pads |
| Color | Blue |
| Diameter (Metric) | 355 mm |
| For Use With (Application) | Spray Cleaning, and Foam Scrubbing |
| For Use With (Equipment) | Rotary and Automatic Floor Machines |
| Style | Round |