Learn More
Cytiva Blotting Paper
Product Code. 11505634 Shop All Cytiva Products
Change view
Click to view available options
Type:
Blotting Paper
Unit Size:
Pack of 50
Product Code. Type unitSize
11505634 Blotting Paper Pack of 50
1 options
Product Code. 11505634
Brand: Cytiva TE26
Only null left
Add to basket
Add to basket
Research laboratory blotting paper for gel electrophoresis applications.
  • For use with mini vertical gel units
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Mini Vertical Electrophoresis Unit
Type Blotting Paper
Quantity 50 Pk.
Length (English) 4.13 in.
Length (Metric) 10.5 cm
Width (English) 3.54 in.
Width (Metric) 9 cm