Learn More
Karl Hecht™ Blood Smear Slides
Product Code. 15844802
Change view
Click to view available options
Product Type:
Undritz blood smear slide
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15844802 Undritz blood smear slide Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15844802 Supplier Karl Hecht™ Supplier No. 40619010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An excellent accessory for the microscopic examination of blood samples.
  • According to Undritz
  • CE marking
  • Clear glass
  • Both sides polished
  • Beveled edges and corners
  • Thickness from 1.6 to 1.7mm
  • Dimensions approx. 40 x 20mm
Specifications
Product Type Undritz blood smear slide
For Use With (Application) Microscopic blood examination