Learn More
Benchmark Scientific™ Block for Deepwell Microplates
Click to view available options
Benchmark Scientific™ Microplate Holder
Description
- Accommodates one deepwell microplate
- Removable lid helps to maintain block temperature
- Notched sides aid in removal of plate
- Fits MultiTherm H and HC Models
Specifications
Autoclavable | Not Autoclavable |
Chamber Material | Aluminum |
Holds | 1 Deepwell Microplate |
Material | Aluminum |
No. of Places | 1 |
Type | Block |