Learn More
Benchmark Scientific™ Block for Deepwell Microplates
Catalog Number 17994402
Click to view available options
Benchmark Scientific™ Microplate Holder
This block holds one deep well microplate. Notched sides make plate removal easy. Made of aluminum with a removable cover. Fits Benchmark MultiTherm.
  • Accommodates one deepwell microplate
  • Removable lid helps to maintain block temperature
  • Notched sides aid in removal of plate
  • Fits MultiTherm H and HC Models
Autoclavable Not Autoclavable
Chamber Material Aluminum
Holds 1 Deepwell Microplate
Material Aluminum
No. of Places 1
Type Block