Description
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Specifications
| Description | Cover Low-Temperature 2 Block |
| Type | Block Cover |
| For Use With (Application) | Low Temperatures |
| No. of Blocks | 2 |