Learn More
BLK Monoclonal Antibody (9D10B7H6, 9D10A8F8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515302
Additional Details : Weight : 0.01000kg
Description
MA5-15302 targets BLK in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15302 immunogen is purified recombinant fragment of BLK expressed in E. Coli.
BLK Monoclonal antibody specifically detects BLK in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
BLK | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P51451 | |
BLK | |
Purified recombinant fragment of BLK expressed in E. Coli. | |
RUO | |
640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
9D10B7H6, 9D10A8F8 | |
Unconjugated | |
BLK | |
B lymphocyte kinase; B lymphoid kinase; B lymphoid tyrosine kinase; Blk; BLK nonreceptor tyrosine kinase; BLK proto-oncogene, Src family tyrosine kinase; MGC10442; MODY11; p55-Blk; tyrosine-protein kinase Blk | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |