Learn More
BRAND™ BLAUBRAND™ Micropipettes
Shop All Brand Products
Click to view available options
:
Volume: 40 μL
Description
- DE-M marking
- Calibrated to contain (TC, In)
- Color code to ISO for clear identification
- CE-marked according to IVD-Directive 98/79 CE
Specifications
Specifications
| Accuracy | ± 0.25% |
| Certifications/Compliance | DE-M |
| Quantity | 250/Pk |
| Packaging Type | Jar |
| Color | 2 x red |
| Volume (Metric) | 40 μL |