Learn More
BRAND™ BLAUBRAND™ Glass Graduated Cylinders, Class A, USP, Certified
Product Code. p-5164470 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
500 mL
2000 mL
Graduations:
0.5 mL
5 mL
20 mL
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Graduations unitSize
15734037 25 mL 0.5 mL Pack of 2
15358286 500 mL 5 mL Pack of 2
15584409 2000 mL 20 mL Each
3 options
Product Code. 15734037
Brand: BRAND™ 32820
Only null left
Add to basket
Add to basket
Class A, USP Certified, calibrated 'To Contain'
  • Manufactured from Borosilicate 3.3 glass, known for its low expansion coefficient and excellent chemical resistance
  • Features a blue enamel print for optimal durability and easy readability
  • Equipped with pour spout and a sturdy hexagonal glass base for convenient handling and stability
  • Available in a full range of ISO sizes from 5 mL to 2000 mL
  • Calibrated 'To Contain'
  • Each unit is supplied with batch number and a USP batch certificate in original packaging for traceability
TRUSTED_SUSTAINABILITY
Specifications
Base Type Hexagonal
Calibration To Contain
Scale Color Blue
Shape Round
Certifications/Compliance DIN EN ISO 4788, USP
Class Class A
Height (Metric) 170 mm
Tolerance Metric ±0.17 mL
Type Graduated Cylinder
Material Borosilicate Glass
Capacity (Metric) 25 mL
Includes USP Batch Certificate
Graduations 0.5 mL
Subdivision 0.5mL