Learn More
BRAND™ Blaubrand™ Eterna™ Class AS Bulb Pipets, with Batch Certificate
Product Code. 15821508 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15821508 10 mL Pack of 6
15811508 5 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15821508 Supplier BRAND™ Supplier No. 230512
  • ONLINE EXCLUSIVE PRICE
    654.00 NOK
    463.72 NOK / Pack of 6
    Save 190.28 NOK
    29% Off
Estimated Shipment: 27-01-2027
Manufactured from AR-GLAS™ Soda-lime glass and complies with DIN EN ISO 648 standards. Delivered with batch number and one batch certificate per packing unit of the manufacturer. Total order delivered from one batch (to the extent possible).
  • Manufactured from AR-GLAS™ Soda-lime glass.
  • DIN EN ISO 648. Calibrated to deliver (TD, Ex).
  • Waiting time 5 seconds.
  • Delivered with batch number and one batch certificate per packing unit of the manufacturer.
  • Total order delivered from one batch (to the extent possible)
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Deliver
Certifications/Compliance DIN EN ISO 648, DIN 12691
Class AS
Material AR-GLAS, Soda Lime Glass
Graduated Yes
Graduation Color Amber
Graduation Type DE-M Marking
Tolerance 0.02 mL
Color Red
Volume (Metric) 10 mL
Type Volumetric Bulb Pipet