Learn More
BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Certificate
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
100 mL
250 mL
Unit Size:
Each
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- Includes polypropylene stopper and DE-M marking Delivered with individual conformity certificate
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Certifications/Compliance | USP individual and USP batch-certificate, DIN EN ISO 1042 |
| Closure Material | Polypropylene |
| Color | Clear |
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Limit of Error | ±0.15 mL |
| Neck Style | Narrow |
| Graduated | Yes |
| Class | A |
| Show More |