Learn More
BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Batch Certificate
Product Code. 15831508 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
200 mL
Unit Size:
Pack of 2
Product Code. 15831508
Brand: BRAND™ 236838
Only null left
Add to basket
Add to basket
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. Calibrated to contain a precise amount of liquid at a specific temperature. Includes polypropylene stopper and DE-M marking. Total order delivered from one batch (to the extent possible).
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • Includes polypropylene stopper and DE-M marking
  • Total order delivered from one batch (to the extent possible)
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042
Closure Material Polypropylene
Color Clear
Capacity (Metric) 5 mL
Material Borosilicate Glass
Limit of Error ±0.04 mL
Neck Style Wide
No. per Case 2 Pack
Graduated Yes
Class A
Closure Type Stopper
Closure Size 10/19