Learn More
BRAND™ Blaubrand™ ETERNA™ Class A USP Volumetric Flasks, with Certificate
Product Code. 15835572 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 15835572
Brand: BRAND™ 956854
Only null left
Add to basket
Add to basket
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. Calibrated to contain a precise amount of liquid at a specific temperature. Includes polypropylene stopper and DE-M marking. Delivered with individual conformity certificate.
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • Includes polypropylene stopper and DE-M marking
  • Delivered with individual USP conformity certificate
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance USP individual and USP batch-certificate, DIN EN ISO 1042
Closure Material Polypropylene
Color Clear
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Limit of Error ±0.6 mL
Neck Style Narrow
Graduated Yes
Class A
Closure Type Stopper
Closure Size 29/32