Learn More
BRAND™ BLAUBRAND™ Class A Volumetric Flask with Three Marks
Product Code. 10522024 Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Description:
Upper/lower mark: 0.07 mL
Upper/lower mark: 0.175 mL
Upper/lower mark: 0.35 mL
Upper/lower mark: 0.7 mL
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
Unit Size:
Each
Product Code. 10522024
Brand: BRAND™ 38210
Only null left
Add to basket
Add to basket
The volumetric flask with three marks is used to check the functioning of a dispenser. The mark in the middle shows the nominal volume, the upper and lower marks show the error limits as specified in the table below. If the error limit is exceeded even with repeated measurements, the dispenser is out of specification.
  • Manufactured from Borosilicate Glass.
  • Beaded rim prevents slippage when suspended by a clamp.
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Batch number and individual certificate included.
  • Dakks calibration certificate available upon request.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN ISO 1042. Calibrated to contain (TC, In). Includes DAkkS-certificate
Description Upper/lower mark: 0.7 mL
Capacity (Metric) 100 mL
Material Borosilicate Glass
Standard Taper Joint NS 14/23
Class A