Learn More
BRAND™ BLAUBRAND™ Class A Volumetric Flask with Three Marks
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Description:
Upper/lower mark: 0.07 mL
Upper/lower mark: 0.175 mL
Upper/lower mark: 0.35 mL
Upper/lower mark: 0.7 mL
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
Unit Size:
Each
Description
- Manufactured from Borosilicate Glass.
- Beaded rim prevents slippage when suspended by a clamp.
- Complies with DIN EN ISO 1042 standards.
- Calibrated to contain (TC, In.)
- Batch number and individual certificate included.
- Dakks calibration certificate available upon request.
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042. Calibrated to contain (TC, In). Includes DAkkS-certificate |
| Description | Upper/lower mark: 0.07 mL |
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass |
| Standard Taper Joint | NS 10/19 |
| Class | A |