Learn More
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark
Shop All Brand Products
Click to view available options
Volume (Metric):
1 mL
6 mL
20 mL
25 mL
Unit Size:
Pack of 12
Pack of 6
Description
- AR-Glas™
- Class 1-mark graduated
- Includes batch certificate of conformity
Specifications
Specifications
| Class | Class AS |
| Material | AR-GLAS |
| Graduation Type | 1-Mark Graduations |
| Includes | Batch Certificate of Conformity |
| Volume (Metric) | 25 mL |
| Type | Bulb Pipet |