Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Compact Burettes
Product Code. 10464342 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10464342 50 mL Each
10397482 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10464342 Supplier BRAND™ Supplier No. 13918
  • ONLINE EXCLUSIVE PRICE
    935.00 NOK
    849.08 NOK / Pack of 1
    Save 85.92 NOK
    9% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Fast to dismantle and easy to clean. BRAND™ Blaubrand™ Borosilicate Glass Compact Burettes are also simple to repair because all individual components replaceable.
  • Includes removable PTFE stopcock
  • Fast to dismantle and easy to clean.
  • Simple repair - all individual components replaceable.
  • Borosilicate glass construction (Boro 3.3 to DIN EN ISO 385.)
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Length (Metric) 800 mm
Material Borosilicate Glass
Graduations 0.1 mL
Includes Removable PTFE Stopcock
Stopcock Material PTFE
Tolerance ±0.05 mL
Class Class AS
Certifications/Compliance DIN EN ISO 385, USP