Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Product Code. 10693371 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10693371 5 mL Each
10663561 10 mL Each
10748171 50 mL Each
10747981 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10693371 Supplier BRAND™ Supplier No. 43305
  • ONLINE EXCLUSIVE PRICE
    600.00 NOK
    554.51 NOK / Pack of 1
    Save 45.49 NOK
    7% Off
View all promotions
Estimated Shipment: 02-12-2026
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
  • Calibrated density bottle made with Borosilicate glass
  • DIN ISO 3507, type Gay-Lussac
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • The volume in cm³ is specified to a precision of 3 decimal places
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 mL
Material Borosilicate Glass
Product Type Calibrated Density Bottle