Learn More
BRAND™ BLAUBRAND™ Automatic Burette with Lateral Stopcock
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
Description
- Calibrated to deliver (TD, Ex)
- Schellbach stripe
- With needle-valve stopcock with PTFE spindle
- Automatic zeroing
- 2000 mL soda-lime glass bottle is sold separately (see 126965, BRAND™ BISTABIL™ Bottles for Automatic Burettes)
- Delivered with batch number and enclosed one batch certificate per pack
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Graduations | 0.1 mL |
| Includes | Blue Schellbach stripe |
| Stopcock | Lateral safety stopcock |
| Warranty | 12 months |
| Type | Buret |