Learn More
BRAND™ BLAUBRAND™ Automatic Burette with Lateral Stopcock
Product Code. 10418952 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10418952 25 mL Each
11956129 10 mL Each
10673202 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10418952 Supplier BRAND™ Supplier No. 22522
  • ONLINE EXCLUSIVE PRICE
    3035.00 NOK
    2757.91 NOK / Pack of 1
    Save 277.09 NOK
    9% Off
Estimated Shipment: 10-11-2026
BLAUBRAND™ Automatic Burette includes a lateral stopcock and blue Schellbach stripe.
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • With needle-valve stopcock with PTFE spindle
  • Automatic zeroing
  • 2000 mL soda-lime glass bottle is sold separately (see 126965, BRAND™ BISTABIL™ Bottles for Automatic Burettes)
  • Delivered with batch number and enclosed one batch certificate per pack
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Graduations 0.05 mL
Includes Blue Schellbach stripe
Stopcock Lateral safety stopcock
Warranty 12 months
Type Buret