Learn More
BRAND™ BLAUBRAND™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper, with Certificate
Product Code. 15885552 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
500 mL
Unit Size:
Each
Product Code. 15885552
Brand: BRAND™ 937452
Only null left
Add to basket
Add to basket
Manufactured from Amber Borosilicate Glass and compllies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper protects light-sensitive liquids from deterioration. Delivered with individual conformity certificate.
  • Manufactured from Amber Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Includes amber glass stopper, interchangeable with polypropylene stopper.
  • Dark amber flask protects light-sensitive liquids from deterioration.
  • Batch number and certificate enclosed with packing.
  • Delivered with individual conformity certificate.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042. DAkkS calibration certificate upon request.
Closure Material Glass
Color Amber
Capacity (Metric) 500 mL
Material Borosilicate Glass
Limit of Error ±0.25 mL
Neck Style Narrow
No. per Case 2 Piece
Tolerance ±0.25 mL
Class A
Closure Size 19/26