Learn More
BRAND™ Blaubrand™ Amber Borosilicate Glass Class A Volumetric Flask, with Certificate
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
500 mL
1000 mL
Unit Size:
Each
Description
- Manufactured from Amber Borosilicate Glass.
- Complies with DIN EN ISO 1042 standards.
- Calibrated to contain (TC, In.)
- The dark glass protects against deterioration of light-sensitive liquids.
- Includes polypropylene stopper, but is interchangeable with amber glass stopper.
- Batch number and certificate included. Delivered with individual conformity certificate.
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Certifications/Compliance | DIN EN ISO 1042. DAkkS calibration and USP certificates upon request. |
| Closure Material | Polypropylene |
| Color | Amber |
| Capacity (Metric) | 200 mL |
| Material | Borosilicate Glass |
| Limit of Error | ±0.15 mL |
| Neck Style | Narrow |
| No. per Case | 2 Piece |
| Tolerance | ±0.15 mL |
| Show More |