Learn More
Brand™ Blaubrand™ Amber Borosilicate Glass Class A USP Volumetric Flask With Glass Stopper
Product Code. 15252845 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
100 mL
Closure Size:
10/16
14/23
Product Type:
Volumetric Flask
Limit of Error:
±0.03 mL
±0.08 mL
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Product Type Limit of Error unitSize
15252845 25 mL 10/16 Volumetric Flask ±0.03 mL Pack of 2
15272845 100 mL 14/23 Volumetric Flask ±0.08 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15252845 Supplier Brand™ Supplier No. 37464
Only null left
Add to basket
Add to basket
Manufactured from Amber Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Amber Borosilicate Glass Class A USP Volumetric Flask with Glass Stopper protects light-sensitive liquids from deterioration. Glass stopper is Interchangeable with polypropylene stopper. USP individual certificate available upon request.
  • Manufactured from Amber Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Includes amber glass stopper, interchangeable with polypropylene stopper.
  • Dark amber flask protects light-sensitive liquids from deterioration.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042, USP individual certificate by request
Closure Material Glass
Color Amber
Capacity (Metric) 25 mL
Material Borosilicate Glass
Limit of Error ±0.03 mL
No. per Case 2 Piece
Product Type Volumetric Flask
Tolerance ±0.03 mL
Class A
Closure Size 10/16