Learn More
Brady™ Blank Label Core, 76 mm Diameter
Click to view available options
Unit Size:
Each
Description
One blank label core for use with Brady™ printers.
Specifications
Specifications
| For Use With (Equipment) | Brady Label Printers |
| Width (English) | 4.3 in. |
| Width (Metric) | 110 mm |
| Diameter (English) | 3 in. |
| Diameter (Metric) | 76.2 mm |
| Certifications/Compliance | 2011/65/EU and 2015/863 |