Learn More
Scientific Industries SI™ Blank Foam Inserts
Product Code. 15587315 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie 2 Recessed Platform
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15587315 Vortex-Genie 2 Recessed Platform Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15587315 Supplier Scientific Industries SI™ Supplier No. 83041432
  • ONLINE EXCLUSIVE PRICE
    329.70 NOK
    300.31 NOK / Pack of 1
    Save 29.39 NOK
    8% Off
Estimated Shipment: 05-11-2026
Allows users to accommodate odd-shaped vessels. Scientific Industries SI™ Blank Foam Inserts are made of high-density foam that fits into a Vortex-Genie 2 Recessed Platform.
  • Constructed of high-density foam
  • Customizable for odd-shaped vessels.
  • Fits into the Recessed Platform
  • For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
  • One insert is included in the Large Sample Set
TRUSTED_SUSTAINABILITY
Specifications
Material High-density foam
For Use With (Equipment) Vortex-Genie 2 Recessed Platform