Learn More
R&D Systems™ Blank Comp-Bead Particles
Product Code. 18327516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18327516 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18327516 Supplier R&D Systems™ Supplier No. NBP300500
 Restricted Item
Estimated Shipment: 08-12-2026
A Novus product, part of R&D Systems. The Blank Compensation Beads serve as a negative control for setting compensation in multicolor flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C in the dark. Do not freeze.
Format 0.016 M PBS (pH 7.4), 0.2% BSA
Preservative 0.02% Sodium Azide
Quantity 5 mL
For Use With (Application) Flow Cytometry
Product Line Novus
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.