Learn More
IKA™ Blade Stirrer
Product Code. 15553101 Shop All IKA Products
Change view
Click to view available options
Length (Metric):
359 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
15553101 359 mm Each
1 options
Product Code. 15553101
Brand: IKA™ 0020001206
Only null left
Add to basket
Add to basket
Overhead, multi-blade stirrer element is used to mix material from top and bottom at medium to high speeds.
  • Generates a radial flow
  • For drawing the material to be mixed from the top and bottom
  • For aerating low viscosity media and homogenizing solids in liquids
  • Use at medium to high speeds
TRUSTED_SUSTAINABILITY
Specifications
Description Model R 3004
Diameter (English) Fan 1.2 in.
Diameter (English) Shaft 0.3 in.
Diameter (Metric) Fan 30 mm
Diameter (Metric) Shaft 8 mm
For Use With (Application) Aerating low viscosity media and homogenizing solids in liquids
Length (English) 14 in.
Length (Metric) 359 mm
Type Blade Impeller