Learn More
Cole-Parmer™ Viton™ Black Tubing
Product Code. 11775399
Change view
Click to view available options
Diameter (Metric) Outer:
11.2 mm
12.8 mm
22.4 mm
Thickness (Metric):
1.6 mm
3.2 mm
Maximum Pressure:
17 psi
18 psi
20 psi
Unit Size:
7.5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Thickness (Metric) Maximum Pressure unitSize
11775399 11.2 mm 1.6 mm 20 psi 7.5m
11841920 12.8 mm 1.6 mm 18 psi 7.5m
11851920 22.4 mm 3.2 mm 17 psi 7.5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11775399 Supplier Cole-Parmer™ Supplier No. 0643405
Only null left
Add to basket
Add to basket
A strong and durable option for tubing in the laboratory, designed for use with acid and solvent transfer, as well as in high-temperature work.
  • Opaque black in color
  • Tubing is manufactured acc. to GMP
  • Sterilization is not recommended
  • Temperature range from −32° to +204°C
  • Provided in lengths of 25 feet/pack
Specifications
Certifications/Compliance GMP
Diameter (Metric) Inner 8 mm
Material Viton
Diameter (Metric) Outer 11.2 mm
Overall Length (Metric) 7.62 m
Thickness (Metric) 1.6 mm
For Use With (Application) Acid and solvent transfer, high-temperature work
Min. Operating Temperature (Metric) -32°C
Packaging Quantity 1 Each
Sterility Sterile
Sterilization Method Not recommended
Color Black
Hardness 75 (Shore A)
Maximum Pressure 20 psi
Max. Operating Temperature (Metric) 204°C