Learn More
Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly
Product Code. 14863572 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
5 mL
10 mL
Unit Size:
Pack of 40
Product Code. 14863572
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides for accurate, reliable dispensing of various samples. Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly are manufactured from clear Type 1 neutral glass.
  • With screw neck
  • With black caps and bulbs
  • Droppers are assembled but not fitted to vials
  • Dropper assembly comprised of clear glass bottle dropper tube fitted to a polypropylene cap with a natural rubber bulb
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 20 mm
Material Borosilicate Glass
Volume (Metric) 5 mL
Neck Style Screw
Thread Style Screw
Closure Material Polypropylene
Closure Type Dropper Screw Cap
Height (Metric) 43 mm
Quantity 40/Pk.
Type Screw Thread Vial