Learn More
- Home
- Products
- Stoppers and Closures
- Bottle Caps
- Kautex™ Black Polypropylene Screw Closures
Kautex™ Black Polypropylene Screw Closures
Product Code. 10697161| Product Code. | For Use With (Equipment) | Diameter (Metric) | Includes | unitSize |
|---|---|---|---|---|
| 10697161 | 50 mL Bottle | 25 mm | w/out Tamper Evident Ring | Each |
| 15921421 | 50 mL Bottle | 25 mm | w/out Tamper Evident Ring | Pack of 100 |
| 15981471 | 100 mL Bottle | 32 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10628271 | 100 mL Bottle | 32 mm | w/out Tamper Evident Ring | Each |
| 15921411 | 50 mL Bottle | 32 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10615331 | 50 mL Bottle | 32 mm | w/out Tamper Evident Ring | Each |
| 10749221 | 100 mL Bottle | 40 mm | w/Tamper Evident Ring | Pack of 100 |
| 15981451 | 100 mL Bottle | 40 mm | w/Tamper Evident Ring | Pack of 100 |
| 10632371 | 100 mL Bottle | 40 mm | w/out Tamper Evident Ring | Each |
| 15931411 | 100 mL Bottle | 40 mm | w/out Tamper Evident Ring | Pack of 100 |
| 15991471 | 250/350 mL Bottle | 45 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10568822 | 250/350 mL Bottle | 45 mm | w/out Tamper Evident Ring | Each |
| 10360811 | 200/300 mL Bottle | 50 mm | w/out Tamper Evident Ring | Each |
| 15941411 | 200/300 mL Bottle | 50 mm | w/out Tamper Evident Ring | Pack of 100 |
| 15991451 | 200/300 mL Bottle | 50 mm | w/Tamper Evident Ring | Pack of 100 |
| 15901481 | 500/750/1000 mL Bottle | 54 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10569252 | 500/750/1000 mL Bottle | 54 mm | w/out Tamper Evident Ring | Each |
| 15951411 | 500 mL Bottle | 65 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10129570 | 500 mL Bottle | 65 mm | w/out Tamper Evident Ring | Each |
| 10613822 | 500 mL Bottle | 65 mm | w/Tamper Evident Ring | Each |
| 17888167 | 500 mL Bottle | 65 mm | w/Tamper Evident Ring | Pack of 100 |
| 10483511 | 2000 mL Bottle | 80 mm | w/out Tamper Evident Ring | Each |
| 17898147 | 2000 mL Bottle | 80 mm | w/out Tamper Evident Ring | Pack of 100 |
| 15961411 | 750/1000 mL Bottle | 80 mm | w/out Tamper Evident Ring | Pack of 100 |
| 10087950 | 750/1000 mL Bottle | 80 mm | w/out Tamper Evident Ring | Each |
| 10491771 | 1500/2500/4000 mL Bottle | 80 mm | w/out Tamper Evident Ring | Each |
Product Code. 10697161
Brand: Kautex™ 2000071150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Color | Black |
| Includes | w/out Tamper Evident Ring |
| Material | Polypropylene |
| Thread Size (Metric) | 00 mm |
| Liner Material | Polyethylene Foam Insert |
| Neck Finish | 00 mm |
| Product Type | Screw Cap |
| For Use With (Equipment) | 50 mL Bottle |
| Diameter (Metric) | 25 mm |