Learn More
asecos™ Black PE Drum Chock
Product Code. 17856396 Shop All asecos Products
missing translation for 'orderingAttributeHoverText'
Material:
Polyethylene
missing translation for 'unitSize'
Each
Product Code. 17856396
missing translation for 'mfr': asecos™ 12124
This item requires a price update prior to being ordered.
Request a Price for this item here.
Black PE Drum Chock is ideal for tipping drums.
  • PE black to tip drums.
  • Optimal emptying of the drum possible using a pump.
  • Dimensions W x D x H (mm): 217 x 214 x 97
Specifications
Color Black
Material Polyethylene
Type Poly Drum Cradle
Length (English) 8.54 in.
Length (Metric) 217.00 mm
Width (English) 8.54 in.
Width (Metric) 217.00 mm
Height (English) 3.82 in.
Height (Metric) 97.00 mm
Depth (English) 8.43 in.
Depth (Metric) 214.00 mm
Weight (English) 2.43 lb.
Weight (Metric) 1.1 kg