Learn More
Bel-Art™ Scienceware™ Black Insulated Lab Ice Pans
Product Code. 12358697 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
1 L
4 L
9 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
12358697 1 L Each
12338707 9 L Each
12388697 4 L Each
3 options
Product Code. 12358697
Brand: Bel-Art™ Scienceware™ M188481192
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from foam polymer. Bel Art™ Black Insulated Lab Ice Pans is designed for use with dry ice alcohol slurries and liquid nitrogen.
  • Temperature range: -196°C to 90°C
  • Non-sweating, superior insulation properties
  • Leakproof
  • Chemical resistant
  • Floating tube racks available (float in water, dry ice slurries and liquid nitrogen) For use with dry ice, dry ice alcohol slurries and liquid nitrogen.
Specifications
For Use With (Application) For use with ice, dry ice, dry ice alcohol slurries and liquid nitrogen
Material Foam Polymer
Type Insulated Lab Ice Pan
Color Black
Capacity (Metric) 1 L
Temperature Range -196°C to +90°C