Learn More
Uvex Black Busingess Casual 95112 Low Shoe - Width 11
Product Code. 17207851 Shop All Uvex Products
Click to view available options
Size:
43
Unit Size:
1 pair
Product Code. 17207851
Brand: Uvex 9511243
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Uvex business Casual safety shoes for comfortable protection in almost every situation; protection classes S1 and S1 P
Uvex business Casual 9511.2 S1 P SRC
  • Sporty safety shoe
  • Lightweight and flexible
  • Made with new developed last technology for extended wearing comfort
  • Uvex Climazone technology ensures optimum wearer comfort
  • Breathable leather upper
  • Distance mesh lining provides optimum breathability
  • Removable comfort insole, antistatic with moisture control system and additional shock absorption in heel and front of foot
  • Soft padded collar and tongue
  • Steel toe cap and midsole
  • Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
  • Dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
    • Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Certified with Uvex medicare for individual orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 9511.2
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 48
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Leather
Size 43
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Steel
Disposable Reusable
Certifications/Compliance (Europe) EN ISO 20345:2011
Gender Unisex
Fit Standard width
Model Uvex Business Casual
Latex Free Yes
Waterproof No
EN ISO 20345 S1
Slip Resistance Rating SRC