Learn More
Black/Black Flexible PVC Peri-Pump Tubing, 2-Bridge, 0.76 mm (0.03 in.), 12/pack
Click to view available options
Color:
Black/Black
Material:
PVC
Specifications
Specifications
| Diameter (Metric) Inner | 0.76 mm |
| Material | PVC |
| For Use With | Peristaltic Pumps |
| Color | Black/Black |
| Flexibility | Flexible |
| Quantity | 12/Pk. |