Learn More
UVP BL-15 UV Inspection Lamp Replacement Tube
Product Code. p-2622107
Change view
Click to view available options
Length (Metric):
434.8 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12853848 434.8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12853848 Supplier UVP Supplier No. 34001701
Longwave 365nm BLB UV lamp is ideal for inspection applications in hard to reach places.
Specifications
Wattage 15 W
Wavelength 365 nm
For Use With (Equipment) UVP™ BL-15 UV inspection lamp
Length (English) 17.12 in.
Length (Metric) 434.8 mm
Packaging Quantity 1 Ea.