Learn More
Bisphenol A-2,2',6,6'-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Bisphenol A-2,2',6,6'-d4 |
| CAS | 102438-62-0 |
| Molecular Formula | C15 D4 H12 O2 |
| Quantity | 2 mg |
| Synonym | Phen-3,5-d2-ol, 4,4'-(1-methylethylidene)bis- (9CI), Bisphenol A-2,2',6,6'-d4, Bisphenol A-2,2',6,6'-D4, 4,4'-(1-Methylethylidene)bisphen-3,5-d2-ol |
| InChI Formula | InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i3D,4D,5D,6D |
| SMILES | [2H]c1cc(O)cc([2H])c1C(C)(C)c2c([2H])cc(O)cc2[2H] |
| IUPAC Name | 3,5-dideuterio-4-[2-(2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 232.311 |
| Formula Weight | 232.1401 |