Learn More
Bismuth(III) oxyiodide, 98%, pure, Thermo Scientific™
Product Code. p-7077439 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
15259866 10 g 10g
15269866 50 g 50g
2 options
Product Code. 15259866
Brand: Thermo Scientific Acros 453830100
Request Bulk or Custom Format
Chemical Identifiers
CAS 7787-63-5
Molecular Formula BiIO
Molecular Weight (g/mol) 351.88
InChI Key FCBARUADRQNVFT-UHFFFAOYSA-N
Synonym bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi
PubChem CID 6367235
IUPAC Name oxobismuth;hydroiodide
SMILES O=[Bi].I
Specifications
Quantity 10 g
Assay Percent Range 97.5% min
Linear Formula IBi=O
Merck Index 15,127
Solubility Information Solubility in water: insoluble.
Formula Weight 351.88
Percent Purity 98%
Grade Pure
Chemical Name or Material Bismuth(III) oxyiodide
compliance-icons
Product Identifier
  • Bismuth(III) oxyiodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.