Learn More
Bismuth(III) Chloride, Sigma-Aldrich™
Product Code. 15837920
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15837920 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15837920 Supplier Sigma Aldrich Supplier No. 1124030100
 Restricted Item
Estimated Shipment: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Specifications
Boiling Point 447°C/1013 hPa
Melting Point 230°C to 232°C
CAS 7787-60-2
Quantity 100 g
Density 4.75 g/cm3 at 20°C
MDL Number MFCD00003461
Synonym Bismuth(III) chloride, Bismuth trichloride, Bismuth(III) chloride
Molecular Weight (g/mol) 315.34
Grade Research
Chemical Name or Material Bismuth(III) Chloride