missing translation for 'onlineSavingsMsg'
Learn More

Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL

Product Code. p-4408533
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
11339149 100 mL 100mL
11306339 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 11339149

Brand: Thermo Scientific Alfa Aesar 088054.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-69-9 | Bi | 208.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9 , 7732-18-5
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Quantity 100 mL
Grade AAS Standard
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Bismuth: 0.1%; Water: 94.9%
Chemical Name or Material Bismuth
compliance-icons
Product Identifier
  • Bismuth, AAS standard solution, Specpure, Bi 1000µg/ml
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.