Learn More
Bismarck Brown R; MP Biomedicals
Product Code. 11479001 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479001 100 g 100g
11469001 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11479001 Supplier MP Biomedicals Supplier No. 0215485880
  • ONLINE EXCLUSIVE PRICE
    1095.00 NOK
    1040.35 NOK / 100g
    Save 54.65 NOK
    4% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Chemical Identifiers
CAS 5421-66-9
Molecular Formula C21H26Cl2N8
Molecular Weight (g/mol) 461.395
InChI Key WLKAMFOFXYCYDK-UHFFFAOYSA-N
Synonym C.I. 21010, Basic Brown 4
PubChem CID 79459
IUPAC Name 4-[[3-[(2,4-diamino-5-methylphenyl)diazenyl]-4-methylphenyl]diazenyl]-6-methylbenzene-1,3-diamine;dihydrochloride
SMILES CC1=C(C=C(C=C1)N=NC2=C(C=C(C(=C2)C)N)N)N=NC3=C(C=C(C(=C3)C)N)N.Cl.Cl
Specifications
Assay Percent Range ∽ 60%
Formula Weight 461.4
Quantity 100 g
Chemical Name or Material Bismarck Brown R