Learn More
Thermo Scientific Chemicals Bismarck Brown R
Product Code. 15446569
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446569 100 g 100g
15436569 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15446569 Supplier Thermo Scientific Chemicals Supplier No. J64757.22
 Restricted Item
In Stock
Add to basket
CAS: 5421-66-9 | C21H26Cl2N8 | 461.395 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5421-66-9
Molecular Formula C21H26Cl2N8
Molecular Weight (g/mol) 461.395
MDL Number MFCD00036386
InChI Key WLKAMFOFXYCYDK-UHFFFAOYSA-N
Synonym C.I. 21010; Basic Brown 4
PubChem CID 79459
IUPAC Name 4-[[3-[(2,4-diamino-5-methylphenyl)diazenyl]-4-methylphenyl]diazenyl]-6-methylbenzene-1,3-diamine;dihydrochloride
SMILES CC1=C(C=C(C=C1)N=NC2=C(C=C(C(=C2)C)N)N)N=NC3=C(C=C(C(=C3)C)N)N.Cl.Cl
Specifications
Melting Point 222°C (decomposition)
Beilstein 1829673
Formula Weight 461.39
Odor Odorless
Color Black to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Bismarck Brown R