missing translation for 'onlineSavingsMsg'
Learn More

Bis-Tris, ≥99%, MP Biomedicals™

Product Code. 11410822 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 11410822

Brand: MP Biomedicals 0219454690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym 2, 2-bis(Hydroxymethyl)-2, 2', 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 209.2
Percent Purity ≥99%
Chemical Name or Material Bis-Tris, Cell culture reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.